1-Methyl-3-propylimidazolium iodide ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Methyl-3-n-propylimidazolium iodide, 1-Methyl-3-propyl-1H-imidazol-3-ium iodide
M1440-25G
M1440-5G
TCIAM1440-25GEA
61
EUR
TCIAM1440-25G
TCIAM1440-5G
1-Methyl-3-propylimidazolium iodide ≥97.0% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium iodide
Formula:
C₇H₁₃IN₂ Peso molecolare: 252,1 g/mol |
MDL Number:
MFCD08276385 CAS Number: 119171-18-5 |
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...