Stampa…

1-Ethyl-3-methylimidazolium iodide ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-ethyl-3-methylimidazolium iodide

E0556-25G E0556-5G
TCIAE0556-25GEA 90 EUR
TCIAE0556-25G TCIAE0556-5G
1-Ethyl-3-methylimidazolium iodide ≥98.0% (by titrimetric analysis)
1-Ethyl-3-methylimidazolium iodide
Formula: C₆H₁₁IN₂
Peso molecolare: 238,07 g/mol
MDL Number: MFCD03701101
CAS Number: 35935-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8c8714122bdc48e4d0ddedd2b88c2038abb43f0&vid=e8c8714122bdc48e4d0ddedd2b88c2038abb43f0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystaline
Purity (Potassium iodate Method) min. 98.0 %
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR