5-Fluorosalicylaldehyde ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Fluoro-2-hydroxybenzaldehyde
F0574-25G
F0574-5G
TCIAF0574-25GEA
185
EUR
TCIAF0574-25G
TCIAF0574-5G
5-Fluorosalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Fluorosalicylaldehyde
Formula:
C₇H₅FO₂ Peso molecolare: 140,11 g/mol Punto di ebollizione: 56°C |
MDL Number:
MFCD01090997 CAS Number: 347-54-6 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...