Isobutyl isobutyrate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₁₆O₂ Punto di ebollizione: 147°C Flash Pt: 42°C |
MDL Number:
MFCD00008916 CAS Number: 97-85-8 EINECS: 202-612-5 UN: 2528 ADR: 3,III Merck Index: 14,05145 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...