Stampa…

Isobutirrile cloruro ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

I0115-25G I0115-500G I0115-100G
TCIAI0115-25GEA 16 EUR
TCIAI0115-25G TCIAI0115-500G TCIAI0115-100G
Isobutirrile cloruro ≥98.0% (by GC, titration analysis)
Isobutirrile cloruro
Formula: C₄H₇ClO
Peso molecolare: 106.55 g/mol
Punto di ebollizione: 93°C
Flash Pt: 9°C
MDL Number: MFCD00000717
CAS Number: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6a35fac410847737fbbe7162eefdf387885f7a4&vid=b6a35fac410847737fbbe7162eefdf387885f7a4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR