Stampa…

(R)-(-)-2-Aminobutyric acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: D(-)-2-Aminobutyric acid, H-D-Abu-OH

A1377-1G A1377-5G
TCIAA1377-1GEA 17 EUR
TCIAA1377-1G TCIAA1377-5G
(R)-(-)-2-Aminobutyric acid ≥98.0% (by titrimetric analysis)
(R)-(-)-2-Aminobutyric acid
Formula: C₄H₉NO₂
Peso molecolare: 103,12 g/mol
MDL Number: MFCD00064414
CAS Number: 2623-91-8
EINECS: 220-084-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4f6c62ba56159eb8026fa17d9329abd92802746&vid=c4f6c62ba56159eb8026fa17d9329abd92802746&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -20.0 to -22.0 deg (C=5, HCl (2+1))
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR