Stampa…

Methylmalonic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Methylmalonic acid

M0568-100G M0568-25G
TCIAM0568-100GEA 153 EUR
TCIAM0568-100G TCIAM0568-25G
Methylmalonic acid ≥98.0% (by titrimetric analysis)
Methylmalonic acid
Formula: C₄H₆O₄
MDL Number: MFCD00002656
CAS Number: 516-05-2
EINECS: 208-219-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5d458474f17a710ab129a28a676a0ebfbf391c&vid=4d5d458474f17a710ab129a28a676a0ebfbf391c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR