Ethylmalonic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
E0137-25G
E0137-5G
TCIAE0137-25GEA
280
EUR
TCIAE0137-25G
TCIAE0137-5G
Ethylmalonic acid ≥98.0% (by titrimetric analysis)
Ethylmalonic acid
Formula:
C₅H₈O₄ |
MDL Number:
MFCD00002668 CAS Number: 601-75-2 EINECS: 210-007-2 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 112.0 to 117.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...