Dimethylmalonic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,2-Dimethylmalonic acid
D2606-25G
D2606-5G
TCIAD2606-25GEA
83
EUR
TCIAD2606-25G
TCIAD2606-5G
Dimethylmalonic acid ≥98.0% (by GC, titration analysis)
Dimethylmalonic acid
Formula:
C₅H₈O₄ Peso molecolare: 132,12 g/mol |
MDL Number:
MFCD00004193 CAS Number: 595-46-0 EINECS: 209-867-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...