Stampa…

5-Iodoanthranilic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

I0049-25G I0049-5G
TCIAI0049-25GEA 85 EUR
TCIAI0049-25G TCIAI0049-5G
5-Iodoanthranilic acid ≥98.0% (by titrimetric analysis)
5-Iodoanthranilic acid
Formula: C₇H₆INO₂
Punto di ebollizione: 360 °C (1013 hPa)
Punto di fusione: 234 °C
MDL Number: MFCD00007849
CAS Number: 5326-47-6
EINECS: 226-205-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea39f5223be0ddc353bcc8187fbac57cae24b273&vid=e65442dca8b0dc6edd39b46ece3c3f8b74755ba8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR