3-Iodobenzonitrile ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₇H₄IN Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00079762 CAS Number: 69113-59-3 UN: 3439 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 and le 42.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...