4-Fluoro-2-nitrobenzoic acid ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
F0737-1G
F0737-5G
TCIAF0737-1GEA
47
EUR
TCIAF0737-1G
TCIAF0737-5G
4-Fluoro-2-nitrobenzoic acid ≥98.0% (by GC)
4-Fluoro-2-nitrobenzoic acid
Formula:
C₇H₄FNO₄ Peso molecolare: 185,11 g/mol |
MDL Number:
MFCD00024300 CAS Number: 394-01-4 EINECS: 206-890-9 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...