Stampa…

4-Amino-3-fluorobenzoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A2165-1G A2165-5G
TCIAA2165-1GEA 46 EUR
TCIAA2165-1G TCIAA2165-5G
4-Amino-3-fluorobenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-3-fluorobenzoic acid
Formula: C₇H₆FNO₂
Peso molecolare: 155,13 g/mol
MDL Number: MFCD01660374
CAS Number: 455-87-8
EINECS: 207-250-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcecf57fae6ba98c978996d8f6e1b62f0c90faef&vid=679141a4c846a49ce794fc7e370a98a03e09c17f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR