Stampa…

4-Amino-2,3,5,6-tetrafluorobenzoic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

A0987-1G A0987-5G
TCIAA0987-1GEA 72 EUR
TCIAA0987-1G TCIAA0987-5G
4-Amino-2,3,5,6-tetrafluorobenzoic acid ≥97.0% (by titrimetric analysis)
4-Amino-2,3,5,6-tetrafluorobenzoic acid
Formula: C₇H₃F₄NO₂
Peso molecolare: 209,1 g/mol
MDL Number: MFCD00007647
CAS Number: 944-43-4
EINECS: 213-409-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=545e3355b34e0a1c65cf7d3ebae4c74c7a83721f&vid=6d42a6569872c0024c19d24d09a0f8d869ba4246&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light red to Green powder to crystalline
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR