Stampa…

6-Fluoroanthranilic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Amino-6-fluorobenzoic acid

F0475-1G F0475-5G
TCIAF0475-1GEA 16 EUR
TCIAF0475-1G TCIAF0475-5G
6-Fluoroanthranilic acid ≥98.0% (by GC, titration analysis)
6-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
Punto di ebollizione: 305 °C (1013 hPa)
Punto di fusione: 165…169 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00067781
CAS Number: 434-76-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bac9b9d0e266a77350f4fc2ae9832329a349dd26&vid=9ceeeb69e5b1423cb511466facf8d45601e67e1f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point (Decomposition) 166.0 to 171.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR