Stampa…

5-Fluoro-2-nitrobenzaldehyde ≥98.0% (by GC)

Fornitore: TCI
Attenzione

F0645-25G F0645-5G
TCIAF0645-25GEA 140 EUR
TCIAF0645-25G TCIAF0645-5G
5-Fluoro-2-nitrobenzaldehyde ≥98.0% (by GC)
5-Fluoro-2-nitrobenzaldehyde
Formula: C₇H₄FNO₃
MDL Number: MFCD00153175
CAS Number: 395-81-3
EINECS: 206-903-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e8828c79077466fbebb5c9b63f020fe736e4a39&vid=da5267597344e2e18cf73a175e9db080cf442788&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 96.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR