Stampa…

2-Cloro-5-nitroanilina ≥98.0% (by GC)

Fornitore: TCI
Pericolo

C0403-25G C0403-500G
TCIAC0403-25GEA 16 EUR
TCIAC0403-25G TCIAC0403-500G
2-Cloro-5-nitroanilina ≥98.0% (by GC)
2-Cloro-5-nitroanilina
Formula: C₆H₅ClN₂O₂
Punto di fusione: 119 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007668
CAS Number: 6283-25-6
EINECS: 228-498-7
UN: 2237
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d9d98cf6cc6ee8cafa1a248686fb7f81cd964a5&vid=7628e0f3cca0179bf7db0cae5c93ee62c269de99&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 121.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR