4-Chlorosalicylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Chloro-2-hydroxybenzoic acid
C1282-25G
C1282-500G
TCIAC1282-25GEA
41
EUR
TCIAC1282-25G
TCIAC1282-500G
4-Chlorosalicylic acid ≥98.0% (by GC, titration analysis)
4-Chlorosalicylic acid
Formula:
C₇H₅ClO₃ Peso molecolare: 172,57 g/mol |
MDL Number:
MFCD00002449 CAS Number: 5106-98-9 EINECS: 225-829-7 |
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...