5-Chloro-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
C0224-25G
C0224-250G
TCIAC0224-25GEA
42
EUR
TCIAC0224-25G
TCIAC0224-250G
5-Chloro-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
5-Chloro-2-nitrobenzoic acid
Formula:
C₇H₄ClNO₄ Peso molecolare: 201,57 g/mol Punto di fusione: 135…138 °C Densità: 1,593 g/cm³ (18 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007290 CAS Number: 2516-95-2 EINECS: 219-738-1 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 138.0 to 141.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...