Stampa…

3-Chloro-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C0988-25G C0988-5G
TCIAC0988-25GEA 156 EUR
TCIAC0988-25G TCIAC0988-5G
3-Chloro-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
3-Chloro-2-nitrobenzoic acid
Formula: C₇H₄ClNO₄
Peso molecolare: 201,57 g/mol
Punto di fusione: 239°C
Densità: 1,566 g/cm³ (18 °C)
MDL Number: MFCD00007087
CAS Number: 4771-47-5
EINECS: 225-313-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04d1422325b8accd8c3af1417c7a03ae8a11d426&vid=84eb13870d263fe9119cac98856fa9b02621de16&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR