Stampa…

3,5-Dichloroanthranilic acid ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Amino-3,5-dichlorobenzoic acid

D1475-25G D1475-5G
TCIAD1475-25GEA 147 EUR
TCIAD1475-25G TCIAD1475-5G
3,5-Dichloroanthranilic acid ≥97.0% (by HPLC, titration analysis)
3,5-Dichloroanthranilic acid
Formula: C₇H₅Cl₂NO₂
Punto di ebollizione: 344 °C (1013 hPa)
Punto di fusione: 226…229 °C
MDL Number: MFCD00017088
CAS Number: 2789-92-6
EINECS: 220-520-3
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eabc52da3d56e9ada04365c27e15a4ec24ed701&vid=0aa7089fc4072bea129f97b23c8581f8ace84fc5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR