Stampa…

4-Chloro-3,5-dinitrobenzonitrile ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

C1011-5G C1011-25G
TCIAC1011-5GEA 64 EUR
TCIAC1011-5G TCIAC1011-25G
4-Chloro-3,5-dinitrobenzonitrile ≥98.0% (by GC, titration analysis)
4-Chloro-3,5-dinitrobenzonitrile
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=703f9b7d2faefd8d7f8f10ba733e74d58c52cdc7&vid=703f9b7d2faefd8d7f8f10ba733e74d58c52cdc7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 140.0 to 144.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR