Stampa…

3,5-Dihydroxybenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: alpha-Resorcylic acid

D0570-25G D0570-100G
TCIAD0570-25GEA 19 EUR
TCIAD0570-25G TCIAD0570-100G
3,5-Dihydroxybenzoic acid ≥98.0% (by GC, titration analysis)
3,5-Dihydroxybenzoic acid
Formula: C₇H₆O₄
Peso molecolare: 154,12 g/mol
Punto di fusione: 236…238 °C
Flash Pt: ≥ 200 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002512
CAS Number: 99-10-5
EINECS: 202-730-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d6a2b2744e24796c4c804988b2f9e80b9b8c2f5&vid=5cce46643982ffbd621f8353d7a0cf73793477ea&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR