2,4,5-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
T2627-25G
T2627-5G
TCIAT2627-25GEA
69
EUR
TCIAT2627-25G
TCIAT2627-5G
2,4,5-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,4,5-Trimethoxybenzoic acid
Formula:
C₁₀H₁₂O₅ Peso molecolare: 212,2 g/mol Punto di ebollizione: 300°C Punto di fusione: 142 °C |
MDL Number:
MFCD00002435 CAS Number: 490-64-2 EINECS: 207-715-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...