Stampa…

4-Dodecyloxybenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(Dodecyloxy)benzoic acid

D2859-25G D2859-5G
TCIAD2859-25GEA 133 EUR
TCIAD2859-25G TCIAD2859-5G
4-Dodecyloxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Dodecyloxybenzoic acid
Formula: C₁₉H₃₀O₃
Peso molecolare: 306,45 g/mol
Punto di ebollizione: 420 °C (1013 hPa)
Punto di fusione: 137°C
MDL Number: MFCD00002543
CAS Number: 2312-15-4
EINECS: 219-004-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7644d92a5266398fd90222a510798b35998fecf1&vid=1dd5bb7be28850fc27b0b61593684658cb3236ec&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Mesomorphic range 91 to 145 deg-C
Solubility in N,N-DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR