Stampa…

3,4-Dinitrobenzoic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

D0823-25G D0823-5G
TCIAD0823-25GEA 151 EUR
TCIAD0823-25G TCIAD0823-5G
3,4-Dinitrobenzoic acid ≥98.0% (by titrimetric analysis)
3,4-Dinitrobenzoic acid
Formula: C₇H₄N₂O₆
Peso molecolare: 212,12 g/mol
Punto di fusione: 165,5…166.5 °C
Densità: 1,674 g/cm³ (20 °C)
MDL Number: MFCD00007255
CAS Number: 528-45-0
EINECS: 208-433-9
Merck Index: 14,03275
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db8ff7071533ac99651df6fd1322faf4eb6ccf7e&vid=46dd35ac6eb5e8042f74ace909e6ef6dbba4647f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 164.0 to 167.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR