Stampa…

4-Nitro-m-anisic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 3-Methoxy-4-nitrobenzoic acid

M2083-25G M2083-5G
TCIAM2083-25GEA 106 EUR
TCIAM2083-25G TCIAM2083-5G
4-Nitro-m-anisic acid ≥98.0% (by GC, titration analysis)
4-Nitro-m-anisic acid
Formula: C₈H₇NO₅
Peso molecolare: 197.15 g/mol
Punto di ebollizione: 405
Punto di fusione: 233
MDL Number: MFCD00007353
CAS Number: 5081-36-7
EINECS: 225-793-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a6248e921ed1dd267936847386070be4612fac7&vid=c57ac0f931dbfad61f25f3ad593030df0f4f0442&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 233.0 to 237.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR