Stampa…

4-Amino-3-hydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A1194-25G A1194-5G
TCIAA1194-25GEA 108 EUR
TCIAA1194-25G TCIAA1194-5G
4-Amino-3-hydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-3-hydroxybenzoic acid
Formula: C₇H₇NO₃
Peso molecolare: 153,14 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00017094
CAS Number: 2374-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c27984f272655857dda75a7dc2550fc27d60628&vid=16cc7ac7ffc1ca3ffd2dab58d7f6705288778fc9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR