Stampa…

3-Cyanobenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

C0446-25G C0446-5G
TCIAC0446-25GEA 131 EUR
TCIAC0446-25G TCIAC0446-5G
3-Cyanobenzoic acid ≥98.0% (by GC, titration analysis)
3-Cyanobenzoic acid
Formula: C₈H₅NO₂
Peso molecolare: 147.13 g/mol
Punto di fusione: 219
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002486
CAS Number: 1877-72-1
EINECS: 217-511-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e1650b51ae0619ff98b336821cb27c24128a12c&vid=2e6fea72c18912cf747c25880380e93c51605ba4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 218.0 to 222.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR