2,4,5-Trihydroxybenzaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
T1766-1G
T1766-5G
TCIAT1766-1GEA
129
EUR
TCIAT1766-1G
TCIAT1766-5G
2,4,5-Trihydroxybenzaldehyde ≥98.0% (by GC)
2,4,5-Trihydroxybenzaldehyde
Formula:
C₇H₆O₄ Peso molecolare: 154,12 g/mol Punto di fusione: 228 °C |
MDL Number:
MFCD00016592 CAS Number: 35094-87-2 |
Risultati del test di specificazione
Appearance | Yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...