Stampa…

5-Nitrosalicylaldehyde ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hydroxy-5-nitrobenzaldehyde

N0255-25G N0255-5G
TCIAN0255-25GEA 87 EUR
TCIAN0255-25G TCIAN0255-5G
5-Nitrosalicylaldehyde ≥97.0% (by GC, titration analysis)
5-Nitrosalicylaldehyde
Formula: C₇H₅NO₄
Peso molecolare: 167,12 g/mol
Punto di fusione: 123…126 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007337
CAS Number: 97-51-8
EINECS: 202-587-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3c3885f57cd66608865195ba6ab5a0bc6bf6b29&vid=01aff072f43bdf88513552ef153b69134f512e0d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 125.0 to 128.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR