Acido Vanillico ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₈O₄ Punto di ebollizione: 353 °C (1013 hPa) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002551 CAS Number: 121-34-6 EINECS: 204-466-8 Merck Index: 14,09931 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 210.0 to 215.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...