2,4-Dibromoanisole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,3-Dibromo-4-methoxybenzene, 2,4-Dibromo-1-methoxybenzene
D1863-25G
D1863-250G
TCIAD1863-25GEA
50
EUR
TCIAD1863-25G
TCIAD1863-250G
2,4-Dibromoanisole ≥98.0% (by GC)
2,4-Dibromoanisole
Formula:
C₇H₆Br₂O Peso molecolare: 265,93 g/mol Punto di ebollizione: 101°C Punto di fusione: 60…61 °C |
MDL Number:
MFCD00000079 CAS Number: 21702-84-1 EINECS: 244-536-5 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 64.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...