Stampa…

4-Bromo-2-fluorobenzaldehyde ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B2002-25G B2002-5G
TCIAB2002-25GEA 98 EUR
TCIAB2002-25G TCIAB2002-5G
4-Bromo-2-fluorobenzaldehyde ≥96.0% (by GC)
4-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
Peso molecolare: 203,01 g/mol
MDL Number: MFCD00143261
CAS Number: 57848-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e702d3bf4c741f46f4e55152619265b6adb664f6&vid=f3b7565e8f19249aadc14e4b3fce831d6aca4406&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 61.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR