2,4-Diamino-6-methyl-1,3,5-triazine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,4-Diamino-6-methyl-s-triazine, 6-Methyl-1,3,5-triazine-2,4-diamine, 6-Methyl-1,3,5-triazine-2,4-diyldiamine
D0583-25G
D0583-500G
TCIAD0583-25GEA
16
EUR
TCIAD0583-25G
TCIAD0583-500G
2,4-Diamino-6-methyl-1,3,5-triazine ≥98.0% (by titrimetric analysis)
2,4-Diamino-6-methyl-1,3,5-triazine
Formula:
C₄H₇N₅ Peso molecolare: 125,13 g/mol |
MDL Number:
MFCD00023192 CAS Number: 542-02-9 EINECS: 208-796-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...