Dichloromethylvinylsilane ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₃H₆Cl₂Si Punto di ebollizione: 94°C Flash Pt: 14°C |
MDL Number:
MFCD00000492 CAS Number: 124-70-9 EINECS: 204-710-3 UN: 2985 ADR: 3,II |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...