Fluoranthene ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
F0016-25G
F0016-500G
F0016-100G
TCIAF0016-25GEA
29
EUR
TCIAF0016-25G
TCIAF0016-500G
TCIAF0016-100G
Fluoranthene ≥98.0% (by GC)
Fluoranthene
Formula:
C₁₆H₁₀ Peso molecolare: 202.26 g/mol Punto di ebollizione: 384°C |
MDL Number:
MFCD00001184 CAS Number: 206-44-0 EINECS: 205-912-4 |
|
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...