Stampa…

2,4-Difluorophenylhydrazinium chloride ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,4-Difluorophenylhydrazine hydrochloride, 1-(2,4-Difluorophenyl)hydrazine hydrochloride

D2698-25G D2698-5G
TCIAD2698-25GEA 122 EUR
TCIAD2698-25G TCIAD2698-5G
2,4-Difluorophenylhydrazinium chloride ≥97.0% (by titrimetric analysis)
2,4-Difluorophenylhydrazinium chloride
Formula: C₆H₇ClF₂N₂
Peso molecolare: 180,58 g/mol
MDL Number: MFCD00013383
CAS Number: 51523-79-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc81e18276125ec55f0e593878d58fff4e4e36f&vid=9cc81e18276125ec55f0e593878d58fff4e4e36f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (Potassium iodate Method) min. 97.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR