2-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Chlorophenylhydrazine hydrochloride, (2-Chlorophenyl)hydrazine hydrochloride, 1-(2-Chlorophenyl)hydrazine hydrochloride
C0826-100G
C0826-25G
TCIAC0826-100GEA
316
EUR
TCIAC0826-100G
TCIAC0826-25G
2-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC)
2-Chlorophenylhydrazinium chloride
Formula:
C₆H₈Cl₂N₂ Peso molecolare: 179,05 g/mol |
MDL Number:
MFCD00012928 CAS Number: 41052-75-9 EINECS: 255-194-1 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Potassium iodate Method) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...