2-Bromo-5-chlorobenzaldehyde ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B2955-25G
B2955-5G
TCIAB2955-25GEA
91
EUR
TCIAB2955-25G
TCIAB2955-5G
2-Bromo-5-chlorobenzaldehyde ≥97.0% (by GC)
2-Bromo-5-chlorobenzaldehyde
Formula:
C₇H₄BrClO |
MDL Number:
MFCD00462870 CAS Number: 174265-12-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...