Stampa…

2-Amino-4-chlorothiophenol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Amino-4-chlorobenzenethiol, 2-Amino-4-chlorophenyl mercaptan, 5-Chloro-2-mercaptoaniline

A1625-25G A1625-5G
TCIAA1625-25GEA 146 EUR
TCIAA1625-25G TCIAA1625-5G
2-Amino-4-chlorothiophenol ≥98.0% (by GC, titration analysis)
2-Amino-4-chlorothiophenol
Formula: C₆H₆ClNS
Peso molecolare: 159,64 g/mol
MDL Number: MFCD00792528
CAS Number: 1004-00-8
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=618b85d6171b2d6cbd5679e492c0ed47c71012f3&vid=618b85d6171b2d6cbd5679e492c0ed47c71012f3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Green powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 52.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR