Stampa…

1-Bromo-2,4-dichlorobenzene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,4-Dichlorobromobenzene

B2187-25G B2187-5G
TCIAB2187-25GEA 208 EUR
TCIAB2187-25G TCIAB2187-5G
1-Bromo-2,4-dichlorobenzene ≥98.0% (by GC)
1-Bromo-2,4-dichlorobenzene
Formula: C₆H₃BrCl₂
Peso molecolare: 225,9 g/mol
MDL Number: MFCD00018333
CAS Number: 1193-72-2
EINECS: 214-778-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73793d50176160a408345b4025460c1e1ea7a119&vid=e82ca6a34acc79f71666c1e1e601a48f2464a3fe&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR