4-tert-Amylphenol ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₁H₁₆O |
MDL Number:
MFCD00002369 CAS Number: 80-46-6 EINECS: 201-280-9 UN: 2430 ADR: 8,III Merck Index: 14,07142 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...