6-Methoxysalicylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Hydroxy-6-methoxybenzoic acid, 6-Hydroxy-o-anisic acid
M1941-5G
M1941-1G
TCIAM1941-5GEA
42
EUR
TCIAM1941-5G
TCIAM1941-1G
6-Methoxysalicylic acid ≥98.0% (by GC, titration analysis)
6-Methoxysalicylic acid
Formula:
C₈H₈O₄ Peso molecolare: 168,15 g/mol |
MDL Number:
MFCD00674090 CAS Number: 3147-64-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...