Stampa…

Methyl-2-bromo-5-methoxybenzoate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Methyl 2-bromo-5-methoxybenzoate, 2-Bromo-5-methoxybenzoic acid methyl ester

M1934-25G M1934-5G
TCIAM1934-25GEA 113 EUR
TCIAM1934-25G TCIAM1934-5G
Methyl-2-bromo-5-methoxybenzoate ≥98.0% (by GC)
Methyl-2-bromo-5-methoxybenzoate
Formula: C₉H₉BrO₃
Peso molecolare: 245,07 g/mol
Punto di ebollizione: 162°C
MDL Number: MFCD00051594
CAS Number: 35450-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b69f679f2284eadd2c05450d8abacaa7d15bfdd1&vid=b69f679f2284eadd2c05450d8abacaa7d15bfdd1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR