Stampa…

4-Nitrothiophenol ≥95.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Nitrobenzene-1-thiol, 4-Nitrobenzenethiol, 1-Mercapto-4-nitrobenzene, p-Nitrothiophenol

N0146-25G N0146-5G
TCIAN0146-25GEA 186 EUR
TCIAN0146-25G TCIAN0146-5G
4-Nitrothiophenol ≥95.0% (by GC, titration analysis)
4-Nitrothiophenol
Formula: C₆H₅NO₂S
Peso molecolare: 155,18 g/mol
MDL Number: MFCD00007343
CAS Number: 1849-36-1
EINECS: 217-436-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0520fad381f0e7aba63ddef7d738033a302d7376&vid=0520fad381f0e7aba63ddef7d738033a302d7376&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 95.0 %
Purity (Iodometric Titration) min. 95.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR