Methyl heptadecanoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Heptadecanoic acid methyll ester, Methyl Margarate, Margaric acid methyll ester
H0566-25G
H0566-5G
TCIAH0566-25GEA
254
EUR
TCIAH0566-25G
TCIAH0566-5G
Methyl heptadecanoate ≥98.0% (by GC)
Methyl heptadecanoate
Formula:
C₁₈H₃₆O₂ Peso molecolare: 284,48 g/mol |
MDL Number:
MFCD00009001 CAS Number: 1731-92-6 EINECS: 217-055-3 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...