Stampa…

Nonanoyl chloride ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Pelargonoyl chloride, Nonanoylchloride

N0372-25G N0372-100G
TCIAN0372-25GEA 27 EUR
TCIAN0372-25G TCIAN0372-100G
Nonanoyl chloride ≥95.0% (by titrimetric analysis)
Nonanoyl chloride
Formula: C₉H₁₇ClO
Peso molecolare: 176,69 g/mol
Punto di ebollizione: 215°C
Flash Pt: 95°C
MDL Number: MFCD00000768
CAS Number: 764-85-2
EINECS: 212-131-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f170b89758748179a782c13037c2e001e44a2c7&vid=0f170b89758748179a782c13037c2e001e44a2c7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 93.0 %
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR