Ethyl-p-anisate ≥99.0% (by GC)
Fornitore: TCI
Formula:
C₁₀H₁₂O₃ Flash Pt: 138°C |
MDL Number:
MFCD00015151 CAS Number: 94-30-4 EINECS: 202-320-8 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...