Methyl-o-anisate ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Methoxybenzoic acid methyl ester, Methyl 2-methoxybenzoate, o-Anisic acid methyl ester, 2-Anisic acid methyl ester
A0483-25ML
A0483-500ML
A0483-100ML
TCIAA0483-25MLEA
22
EUR
TCIAA0483-25ML
TCIAA0483-500ML
TCIAA0483-100ML
Methyl-o-anisate ≥99.0% (by GC)
Methyl-o-anisate
Formula:
C₉H₁₀O₃ Peso molecolare: 166,18 g/mol Punto di ebollizione: 228°C |
MDL Number:
MFCD00008423 CAS Number: 606-45-1 EINECS: 210-118-6 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...